missing translation for 'onlineSavingsMsg'
Learn More

2,4-Bis(trifluoromethyl)benzyl bromide, 97%

Product Code. 11476766
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11476766 1 g 1g
11486766 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11476766 Supplier Thermo Scientific Alfa Aesar Supplier No. B23910.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 140690-56-8 | C9H5BrF6 | 307.033 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140690-56-8
Molecular Formula C9H5BrF6
Molecular Weight (g/mol) 307.033
MDL Number MFCD00010306
InChI Key SWFFFUJOWAJJCH-UHFFFAOYSA-N
Synonym 2,4-bis trifluoromethyl benzyl bromide, 1-bromomethyl-2,4-bis trifluoromethyl benzene, 2,4-bis trifluoromethyl benzylbromide, benzene, 1-bromomethyl-2,4-bis trifluoromethyl, 2,4-bis trifluoromethyl-1-bromomethyl benzene, pubchem4879, acmc-1c0tm, 2,4-bis-trifluoromethyl-benzylbromide, 2,4-bis-trifluoromethyl benzyl bromide, 2,4-bis-trifluoromethyl-benzyl bromide
PubChem CID 518871
IUPAC Name 1-(bromomethyl)-2,4-bis(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1C(F)(F)F)C(F)(F)F)CBr

Specifications

Density 1.637
Boiling Point 112°C (0.2 mmHg)
Flash Point 92°C (197°F)
Refractive Index 1.4511
Quantity 1 g
UN Number UN3265
Beilstein 7995492
Formula Weight 307.04
Percent Purity 97%
Chemical Name or Material 2,4-Bis(trifluoromethyl)benzyl bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.