missing translation for 'onlineSavingsMsg'
Learn More

2,4-Bis(trifluoromethyl)benzoic acid, 98%, Thermo Scientific™

Product Code. 10513611 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10513611

Brand: Thermo Scientific Acros 311730010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 32890-87-2
CAS Min % 97.5
CAS Max % 100.0
Melting Point 109.0°C to 113.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C9H4F6O2
Linear Formula (CF3)2C6H3CO2H
MDL Number MFCD00042496
Quantity 1 g
Synonym 2,4-bis trifluoromethyl benzoic acid, 2,4-bis-trifluoromethylbenzoic acid, 2,4-di trifluoromethyl benzoic acid, benzoic acid, 2,4-bis trifluoromethyl, 2,4-bis-trifluoromethyl-benzoic acid, maybridge4_003283, acmc-20aoeb, pubchem8491, rarechem al bo 0512
Solubility Information Solubility in water: slightly soluble
InChI Key JTOIZLCQNWWDCN-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1C(F)(F)F)C(F)(F)F)C(=O)O
IUPAC Name 2,4-bis(trifluoromethyl)benzoic acid
Molecular Weight (g/mol) 258.12
PubChem CID 141752
Formula Weight 258.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4-Bis(trifluoromethyl)benzoic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Bis(trifluoromethyl)benzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 2,4-Bis(trifluoromethyl)benzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.