missing translation for 'onlineSavingsMsg'
Learn More

2,4-Bis(trifluoromethyl)benzeneboronic acid, 97%

Product Code. 11354925
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354925 1 g 1g
11364925 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11354925 Supplier Thermo Scientific Alfa Aesar Supplier No. L18161.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 153254-09-2 | C8H5BF6O2 | 257.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153254-09-2
Molecular Formula C8H5BF6O2
Molecular Weight (g/mol) 257.93
MDL Number MFCD01631349
InChI Key WLYPBMBWKYALCG-UHFFFAOYSA-N
Synonym 2,4-bis trifluoromethyl phenylboronic acid, 2,4-bis trifluoromethyl benzeneboronic acid, 2,4-bis trifluoromethyl phenylboronicacid, 2,4-bis trifluoromethyl phenyl boronic acid, boronic acid, 2,4-bis trifluoromethyl phenyl, 2,4-bis-trifluoromethylphenylboronic acid, pubchem5262, acmc-209d8w, 2,4-bis trifluoromethyl phenyl, 2,4-bistrifluoromethylbenzeneboronic acid
PubChem CID 2782667
IUPAC Name [2,4-bis(trifluoromethyl)phenyl]boronic acid
SMILES OB(O)C1=CC=C(C=C1C(F)(F)F)C(F)(F)F

Specifications

Quantity 1 g
Formula Weight 257.93
Percent Purity 97%
Chemical Name or Material 2,4-Bis(trifluoromethyl)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.