missing translation for 'onlineSavingsMsg'
Learn More

2,4-Bis(trifluoromethyl)aniline, 97%, Thermo Scientific™

Product Code. 11409007
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11409007

Brand: Thermo Scientific Alfa Aesar B20595.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 367-71-5
Molecular Formula C8H5F6N
Molecular Weight (g/mol) 229.125
MDL Number MFCD00236209
InChI Key UIWVOUBVGBJRNP-UHFFFAOYSA-N
Synonym 2,4-bis trifluoromethyl aniline, 2,4-ditrifluoromethylaniline, 2,4-bis-trifluoromethylaniline, 2,4-bis-trifluoromethyl-phenylamine, benzenamine,2,4-bis trifluoromethyl, benzenamine, 2,4-bis trifluoromethyl, 2,4-bis trifluoromethyl benzenamine, 2,4-bis trifluoromethyl phenylamine, pubchem7127, acmc-1cr1l
PubChem CID 602856
IUPAC Name 2,4-bis(trifluoromethyl)aniline
SMILES C1=CC(=C(C=C1C(F)(F)F)C(F)(F)F)N

Specifications

Boiling Point 50°C (0.4 mmHg)
Odor Recognizable
Refractive Index 1.4372
Quantity 1 g
UN Number UN2810
Beilstein 3319910
Formula Weight 229.13
Percent Purity 97%
Chemical Name or Material 2,4-Bis(trifluoromethyl)aniline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.