missing translation for 'onlineSavingsMsg'
Learn More

2,4-Bis[(furan-2-ylmethyl)amino]-5-sulphamoylbenzoic Acid, Mikromol™

Product Code. 17952496
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17952496 100 mg 100mg
17911196 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17952496 Supplier Mikromol Supplier No. MM0014.04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2,4-Bis[(furan-2-ylmethyl)amino]-5-sulphamoylbenzoic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4-Bis[(furan-2-ylmethyl)amino]-5-sulphamoylbenzoic Acid
CAS 5046-19-5
Analyte or Component Names 2,4-Bis[(furan-2-ylmethyl)amino]-5-sulphamoylbenzoic Acid
Shipping Condition Room Temperature
Molecular Formula C17 H17 N3 O6 S
Certifications/Compliance ISO 17025
Synonym 2,4-Bis[(furan-2-ylmethyl)amino]-5-sulphamoylbenzoic Acid, Furosemide Imp. D (EP), Furosemide Impurity D
InChI Formula InChI=1S/C17H17N3O6S/c18-27(23,24)16-7-13(17(21)22)14(19-9-11-3-1-5-25-11)8-15(16)20-10-12-4-2-6-26-12/h1-8,19-20H,9-10H2,(H,21,22)(H2,18,23,24)
SMILES NS(=O)(=O)c1cc(C(=O)O)c(NCc2occc2)cc1NCc3occc3
IUPAC Name 2,4-bis(furan-2-ylmethylamino)-5-sulfamoylbenzoic acid
Molecular Weight (g/mol) 391.4
Quantity 100 mg
Formula Weight 391.0838 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.