missing translation for 'onlineSavingsMsg'
Learn More

2,4-Bis(chloromethyl)mesitylene, 98%

Product Code. 11491478
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11491478

Brand: Thermo Scientific Alfa Aesar A13838.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1585-17-7 | C11H14Cl2 | 217.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1585-17-7
Molecular Formula C11H14Cl2
Molecular Weight (g/mol) 217.13
MDL Number MFCD00013681
InChI Key JMNDJDCLOQRCJV-UHFFFAOYSA-N
Synonym 2,4-bis chloromethyl-1,3,5-trimethylbenzene, 2,4-bis chloromethyl mesitylene, benzene, 2,4-bis chloromethyl-1,3,5-trimethyl, 2,4-di chloromethyl-1,3,5-trimethylbenzene, 2,4-bis chloromethylmesitylene, acmc-209dhz, bis-chloromethyl mesitylene, bis-chloromethyl-mesitylene, jmndjdcloqrcjv-uhfffaoysa, 2,4-bis-chloromethyl mesitylene
PubChem CID 137095
IUPAC Name 2,4-bis(chloromethyl)-1,3,5-trimethylbenzene
SMILES CC1=CC(C)=C(CCl)C(C)=C1CCl

Specifications

Melting Point 104°C to 107°C
Boiling Point 180°C (20 mmHg)
Quantity 250 g
UN Number UN3261
Beilstein 2520798
Formula Weight 217.14
Percent Purity 98%
Chemical Name or Material 2,4-Bis(chloromethyl)mesitylene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.