missing translation for 'onlineSavingsMsg'
Learn More

2,4'-Bipyridine, 97%

Product Code. 15493667
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493667 250 mg 250mg
11367139 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15493667 Supplier Thermo Scientific Alfa Aesar Supplier No. H27366.MD
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 581-47-5 | C10H8N2 | 156.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 581-47-5
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.188
MDL Number MFCD00006217
InChI Key RMHQDKYZXJVCME-UHFFFAOYSA-N
Synonym 2,4'-bipyridine, 2,4'-dipyridyl, 4-pyridylpyridine, 2,4'-bipyridyl, 2,4-bipyridyl, 2,4'-dipyridine, unii-c6sn6ru851, ccris 3428, 2-4-pyridyl pyridine, 4-pyridyl-2-pyridine
PubChem CID 68488
IUPAC Name 2-pyridin-4-ylpyridine
SMILES C1=CC=NC(=C1)C2=CC=NC=C2

Specifications

Melting Point 59°C to 63°C
Boiling Point 281°C
Quantity 250 mg
UN Number UN2811
Formula Weight 156.19
Percent Purity 97%
Chemical Name or Material 2,4'-Bipyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.