missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Aminophenylthio)acetic Acid, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | 2-(4-Aminophenylthio)acetic Acid |
| CAS | 104-18-7 |
| Molecular Formula | C8H9NO2S |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C8H9NO2S/c9-6-1-3-7(4-2-6)12-5-8(10)11/h1-4H,5,9H2,(H,10,11) |
| SMILES | Nc1ccc(SCC(=O)O)cc1 |
| IUPAC Name | 2-(4-aminophenyl)sulfanylacetic acid |
| Molecular Weight (g/mol) | 183.23 |
| Formula Weight | 183.0354 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction