missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Aminophenyl)-4-(4-pyridyl)thiazole, 97%, Thermo Scientific™

Catalog Number 15458867
Click to view available options
Quantity:
250 mg
5 g
This item is not returnable. View return policy
1153000-38-4
C14H11N3S
253.323
MFCD12114461
BNYZGNWCDWUMSU-UHFFFAOYSA-N
2-4-aminophenyl-4-4-pyridyl thiazole, 4-4-pyridin-4-yl-1,3-thiazol-2-yl aniline
43667817
4-(4-pyridin-4-yl-1,3-thiazol-2-yl)aniline
C1=CC(=CC=C1C2=NC(=CS2)C3=CC=NC=C3)N
This item is not returnable. View return policy
1153000-38-4
C14H11N3S
253.323
MFCD12114461
BNYZGNWCDWUMSU-UHFFFAOYSA-N
2-4-aminophenyl-4-4-pyridyl thiazole, 4-4-pyridin-4-yl-1,3-thiazol-2-yl aniline
43667817
4-(4-pyridin-4-yl-1,3-thiazol-2-yl)aniline
C1=CC(=CC=C1C2=NC(=CS2)C3=CC=NC=C3)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 253.32
Percent Purity 97%
Chemical Name or Material 2-(4-Aminophenyl)-4-(4-pyridyl)thiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'