missing translation for 'onlineSavingsMsg'
Learn More

2,4,7-Triamino-6-phenylpteridine, 98%

Product Code. 11430521
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430521 100 g 100g
11410521 5 g 5g
11420521 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11430521 Supplier Thermo Scientific Alfa Aesar Supplier No. B20044.22
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 396-01-0 | C12H11N7 | 253.269 g/mol

Weak diuretic with potassium sparing properties that blocks sodium reuptake in the kidneys

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 396-01-0
Molecular Formula C12H11N7
Molecular Weight (g/mol) 253.269
MDL Number MFCD00006708
InChI Key FNYLWPVRPXGIIP-UHFFFAOYSA-N
Synonym triamterene, 2,4,7-triamino-6-phenylpteridine, dyrenium, triamteren, dytac, pterofen, pterophene, triamteril, triteren, ademin
PubChem CID 5546
IUPAC Name 6-phenylpteridine-2,4,7-triamine
SMILES C1=CC=C(C=C1)C2=NC3=C(N=C2N)N=C(N=C3N)N

Specifications

Melting Point >300°C
Quantity 100 g
Beilstein 266723
Merck Index 14,9599
Formula Weight 253.27
Percent Purity 98%
Chemical Name or Material 2,4,7-Triamino-6-phenylpteridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.