missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tris(perfluoroheptyl)-1,3,5-triazine, Mass Spec Std, Thermo Scientific™

Product Code. 11399315
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399315 100 mg 100mg
11389315 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11399315 Supplier Thermo Scientific Alfa Aesar Supplier No. L16678.MC

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,4,6-Tris(perfluoroheptyl)-1,3,5-triazine is used as a mass spectrometry standard.

Solubility
Insoluble in water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21674-38-4
Molecular Formula C24F45N3
Molecular Weight (g/mol) 1185.213
MDL Number MFCD00042439
InChI Key BFAUAMWYFFXWLX-UHFFFAOYSA-N
Synonym 2,4,6-tris pentadecafluoroheptyl-1,3,5-triazine, 2,4,6-tris perfluoroheptyl-1,3,5-triazine, 2,4,6-tris 1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecafluoroheptyl-1,3,5-triazine, tris 1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecafluoroheptyl-1,3,5-triazine, acmc-20aoej, perfluorotriheptyl-s-triazine, tri perfluoroheptyl-s-triazine, tris perfluoroheptyl-s-triazine, tris pentadecafluoroheptyl-1,3,5-triazine, 1,3,5-triazine, 2,4,6-tris pentadecafluoroheptyl
PubChem CID 550641
IUPAC Name 2,4,6-tris(1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecafluoroheptyl)-1,3,5-triazine
SMILES C1(=NC(=NC(=N1)C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Melting Point 29°C to 31°C
Boiling Point 150°C (1.5 mmHg)
Quantity 100 mg
Solubility Information Insoluble in water.
Formula Weight 1185.22
Chemical Name or Material 2,4,6-Tris(perfluoroheptyl)-1,3,5-triazine, Mass Spec Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.