missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Tris(perfluoroheptyl)-1,3,5-triazine, Mass Spec Std, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2,4,6-Tris(perfluoroheptyl)-1,3,5-triazine is used as a mass spectrometry standard.
Solubility
Insoluble in water.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
2,4,6-Tris(perfluoroheptyl)-1,3,5-triazine is used as a mass spectrometry standard.
Solubility
Insoluble in water.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 29°C to 31°C |
| Boiling Point | 150°C (1.5 mmHg) |
| Quantity | 500 mg |
| Solubility Information | Insoluble in water. |
| Formula Weight | 1185.22 |
| Chemical Name or Material | 2,4,6-Tris(perfluoroheptyl)-1,3,5-triazine, Mass Spec Standard |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?