missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tris(3-pyridyl)-1,3,5-triazine, 97%

Product Code. 15417617
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15417617 5 g 5g
15407617 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15417617 Supplier Thermo Scientific Alfa Aesar Supplier No. H36052.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 42333-76-6 | C18H12N6 | 312.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42333-76-6
Molecular Formula C18H12N6
Molecular Weight (g/mol) 312.34
MDL Number MFCD23379843
InChI Key UOSYDSDIRAXTTF-UHFFFAOYSA-N
Synonym 2,4,6-tris 3-pyridyl-s-triazine, 2,4,6-tri-3-pyridinyl-1,3,5-triazine, tris pyridin-3-yl-1,3,5-triazine, 2,4,6-tris 3-pyridyl-1,3,5-triazine
PubChem CID 11472427
IUPAC Name 2,4,6-tripyridin-3-yl-1,3,5-triazine
SMILES C1=CC(=CN=C1)C1=NC(=NC(=N1)C1=CN=CC=C1)C1=CN=CC=C1

Specifications

Melting Point >300°C
Color White to Yellow
Quantity 5 g
Formula Weight 312.33
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2,4,6-Tris(3-pyridyl)-1,3,5-triazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.