missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylthiophenol, 97%

Product Code. 11359869
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359869 5 g 5g
11349869 1 g 1g
11369869 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11359869 Supplier Thermo Scientific Alfa Aesar Supplier No. B24798.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1541-10-2 | C9H12S | 152.255 g/mol

2,4,6-Trimethylthiophenol, is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethylthiophenol, is used as pharmaceutical intermediate.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1541-10-2
Molecular Formula C9H12S
Molecular Weight (g/mol) 152.255
MDL Number MFCD00218475
InChI Key PKANQZUPJCMBAK-UHFFFAOYSA-N
Synonym 2,4,6-trimethylthiophenol, 2,4,6-trimethylbenzene-1-thiol, benzenethiol,2,4,6-trimethyl, 2-mercaptomesitylene, 2-sulphanylmesitylene, acmc-20aoem, 2,4,6-trimethyl-benzene-1-thiol
PubChem CID 284449
IUPAC Name 2,4,6-trimethylbenzenethiol
SMILES CC1=CC(=C(C(=C1)C)S)C

Specifications

Quantity 5 g
Sensitivity Air Sensitive
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 152.25
Percent Purity 97%
Chemical Name or Material 2,4,6-Trimethylthiophenol
compliance-icons
Product Identifier
  • 2,4,6-Trimethylthiophenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2,4,6-Trimethylbenzene-1-thiol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.