missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylstyrene, 95%, stab. with 500ppm 4-tert-butylcatechol

Product Code. 11448546
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11448546

Brand: Thermo Scientific Alfa Aesar A15974.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 769-25-5 | C11H14 | 146.233 g/mol

2,4,6-Trimethylstyrene is used as an intermediate to produce other chemicals like 2-Methoxy-1-(2, 4, 6-trimethyl-phenyl)-ethanone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethylstyrene is used as an intermediate to produce other chemicals like 2-Methoxy-1-(2, 4, 6-trimethyl-phenyl)-ethanone.

Solubility
Soluble in methanol.

Notes
It is sensitive to light. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 769-25-5
Molecular Formula C11H14
Molecular Weight (g/mol) 146.233
MDL Number MFCD00008613
InChI Key PDELBHCVXBSVPJ-UHFFFAOYSA-N
Synonym 2,4,6-trimethylstyrene, 1,3,5-trimethyl-2-vinylbenzene, 2-vinylmesitylene, mesitylethylene, vinylmesitylene, styrene, 2,4,6-trimethyl, unii-89fv7yq871, benzene, 2-ethenyl-1,3,5-trimethyl, 2,6-trimethylstyrene, styrene,4,6-trimethyl
PubChem CID 13036
IUPAC Name 2-ethenyl-1,3,5-trimethylbenzene
SMILES CC1=CC(=C(C(=C1)C)C=C)C

Specifications

Density 0.908
Boiling Point 208°C to 209°C
Flash Point 75°C (167°F)
Refractive Index 1.5295
Quantity 5 g
Beilstein 1904365
Solubility Information Soluble in methanol.
Formula Weight 146.23
Percent Purity 95%
Chemical Name or Material 2,4,6-Trimethylstyrene, stabilized with 500ppm 4-tert-butylcatechol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.