missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylphenylhydrazine hydrochloride, 97%

Product Code. p-7058358
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11343096 1 g 1g
11353096 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11343096 Supplier Thermo Scientific Alfa Aesar Supplier No. L10958.03
Add to basket
This item is not returnable. View return policy

CAS: 76195-82-9 | C9H15ClN2 | 186.68 g/mol

2,4,6-Trimethylphenylhydrazine hydrochloride is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethylphenylhydrazine hydrochloride is used as pharmaceutical intermediates.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76195-82-9
Molecular Formula C9H15ClN2
Molecular Weight (g/mol) 186.68
MDL Number MFCD00052268
InChI Key XFGPCULNJGZRTH-UHFFFAOYSA-N
Synonym mesitylhydrazine hydrochloride, 2,4,6-trimethylphenylhydrazine hydrochloride, 1-mesitylhydrazine hydrochloride, 2,4,6-trimethylphenyl hydrazine hydrochloride, hydrazine, 2,4,6-trimethylphenyl-, hydrochloride, mesityl hydrazine hydrochloride
PubChem CID 2774883
IUPAC Name (2,4,6-trimethylphenyl)hydrazine;hydrochloride
SMILES Cl.CC1=CC(C)=C(NN)C(C)=C1

Specifications

Melting Point ∽185°C (decomposition)
Quantity 1 g
UN Number UN2811
Beilstein 3700289
Solubility Information Soluble in water.
Formula Weight 186.69
Percent Purity 97%
Chemical Name or Material 2,4,6-Trimethylphenylhydrazine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.