missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylphenylboronic acid, 95%

Product Code. 15505086 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15505086 1 g 1g
15515086 5 g 5g
15525086 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15505086 Supplier Thermo Scientific Acros Supplier No. 359790010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5980-97-2 | C9H13BO2 | 164.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5980-97-2
Molecular Formula C9H13BO2
Molecular Weight (g/mol) 164.01
InChI Key BZXQRXJJJUZZAJ-UHFFFAOYSA-N
Synonym mesitylboronic acid, 2,4,6-trimethylbenzeneboronic acid, 2-mesityleneboronic acid, 2,4,6-trimethylphenyl boronic acid, 2,4,6-trimethylphenyl boranediol, boronic acid, 2,4,6-trimethylphenyl, 2,4,6-trimethylboronic acid, boronic acid, b-2,4,6-trimethylphenyl, mesityl boronic acid, pubchem7895
PubChem CID 292184
IUPAC Name (2,4,6-trimethylphenyl)boronic acid
SMILES B(C1=C(C=C(C=C1C)C)C)(O)O

Specifications

Melting Point 131.0°C to 136.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 164.01
Percent Purity 95%
Chemical Name or Material 2,4,6-Trimethylphenylboronic acid
compliance-icons
Product Identifier
  • 2,4,6-Trimethylphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.