missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylbenzylamine, 99%

Product Code. 10088851 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10088851

Brand: Thermo Scientific Acros 351180050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 40393-99-5 | C10H16N | 150.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40393-99-5
Molecular Formula C10H16N
Molecular Weight (g/mol) 150.24
MDL Number MFCD03411013
InChI Key DGSRAILDFBJNQI-UHFFFAOYSA-O
Synonym mesitylmethanamine, 2,4,6-trimethylbenzylamine, 2,4,6-trimethylphenyl methanamine, 1-mesitylmethanamine, benzenemethanamine, 2,4,6-trimethyl, 1-2,4,6-trimethylphenyl methanamine, 2,4,6-trimethylphenyl methylamine, 2,4,6-trimethylbenzylamine hydrochloride, 1-mesitylmethylamine, pubchem7408
PubChem CID 458433
SMILES CC1=CC(C)=C(C[NH3+])C(C)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5360 to 1.538
Quantity 5 g
Beilstein 12, IV, 2850
IUPAC Name (2,4,6-trimethylphenyl)methanamine
Formula Weight 149.24
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 4, 6-Trimethylbenzylamine, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2,4,6-Trimethylbenzylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.