missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,4,6-Trimethylbenzyl mercaptan, 97%

Product Code. 11473357
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473357 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11473357 Supplier Alfa Aesar™ Supplier No. A17139.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 21411-42-7
Molecular Formula C10H14S
Molecular Weight (g/mol) 166.282
MDL Number MFCD00173911
InChI Key LJZYEPXPMRRXTA-UHFFFAOYSA-N
Synonym 2,4,6-trimethylbenzyl mercaptan, mesitylmethanethiol, 2,4,6-trimethylphenyl methanethiol, benzenemethanethiol,2,4,6-trimethyl, benzenemethanethiol, 2,4,6-trimethyl, trimethylbenzylmercaptane, acmc-1ch2x, 2,4,6-trimethylbenzylmercaptan, 2,4,6-trimethylphenyl methane-1-thiol
PubChem CID 301953
IUPAC Name (2,4,6-trimethylphenyl)methanethiol
SMILES CC1=CC(=C(C(=C1)C)CS)C

Specifications

Melting Point 46°C to 47°C
Boiling Point 120°C (5 mmHg)
Odor Unpleasant
Quantity 25 g
UN Number UN2811
Sensitivity Air Sensitive
Formula Weight 166.29
Percent Purity 97%
Chemical Name or Material 2,4,6-Trimethylbenzyl mercaptan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.