missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylbenzyl alcohol, 98+%

Product Code. 11351457
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11351457

Brand: Thermo Scientific Alfa Aesar L01952.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4170-90-5 | C10H14O | 150.221 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4170-90-5
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00014422
InChI Key LODDFDHPSIYCTK-UHFFFAOYSA-N
Synonym 2,4,6-trimethylbenzyl alcohol, mesitylmethanol, mesitylcarbinol, 2,4,6-trimethylphenyl methanol, unii-rev3kvt84n, benzenemethanol, 2,4,6-trimethyl, 2,4,6-trimethylbenzylic alcohol, benzyl alcohol, 2,4,6-trimethyl, rev3kvt84n, 2,4,6-trimethylphenyl methan-1-ol
PubChem CID 20139
IUPAC Name (2,4,6-trimethylphenyl)methanol
SMILES CC1=CC(=C(C(=C1)C)CO)C

Specifications

Melting Point 85°C to 89°C
Boiling Point 140°C (15 mmHg)
Quantity 25 g
Beilstein 1862608
Formula Weight 150.22
Percent Purity ≥98%
Chemical Name or Material 2,4,6-Trimethylbenzyl alcohol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.