missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylbenzoyl chloride, 98+%

Product Code. 11427027
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11427027 50 g 50g
11417027 10 g 10g
11437027 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11427027 Supplier Thermo Scientific Alfa Aesar Supplier No. A14407.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 938-18-1 | C10H11ClO | 182.647 g/mol

2,4,6-Trimethylbenzoyl chloride is used, in the presence of pyridine, to protect OH groups as mesitoate esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethylbenzoyl chloride is used, in the presence of pyridine, to protect OH groups as mesitoate esters.

Solubility
Reacts slowly with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature: 2 - 8°C. Air and moisture sensitive. Store under nitrogen. Incompatible with strong oxidizing agent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 938-18-1
Molecular Formula C10H11ClO
Molecular Weight (g/mol) 182.647
MDL Number MFCD00013650
InChI Key UKRQMDIFLKHCRO-UHFFFAOYSA-N
Synonym mesitoyl chloride, benzoyl chloride, 2,4,6-trimethyl, 2,4,6-trimethylbenzoylchloride, 2,4,6-trimethyl-benzoyl chloride, pubchem13234, acmc-209rno, 2-mesitylenecarbonyl chloride, ksc491c5b, 2,4,6trimethylbenzoyl chloride, 2,4,6-trimethylbenzoyl cloride
PubChem CID 97038
IUPAC Name 2,4,6-trimethylbenzoyl chloride
SMILES CC1=CC(=C(C(=C1)C)C(=O)Cl)C

Specifications

Density 1.1
Boiling Point 143°C to 146°C (60 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.529
Quantity 50 g
UN Number UN3265
Beilstein 776108
Sensitivity Moisture sensitive
Solubility Information Reacts slowly with water.
Formula Weight 182.65
Percent Purity ≥98%
Chemical Name or Material 2,4,6-Trimethylbenzoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.