missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylbenzonitrile, 98%

Product Code. 11443854
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11443854 25 g 25g
11433854 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11443854 Supplier Thermo Scientific Alfa Aesar Supplier No. A11882.14
  • ONLINE EXCLUSIVE PRICE
    £115.00
    £103.65 / 25g
    Save £11.35
    9% Off
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2571-52-0 | C10H11N | 145.205 g/mol

It employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2571-52-0
Molecular Formula C10H11N
Molecular Weight (g/mol) 145.205
MDL Number MFCD00016378
InChI Key SNIZBGQMWRHNDY-UHFFFAOYSA-N
Synonym mesitonitrile, benzonitrile, 2,4,6-trimethyl, 2,4,6-trimethyl-benzonitrile, .beta.-isodurylonitrile, 2,4,6-trimethylbenzenecarbonitrile, cyanmesitylen, cyanomesitylene, mesitylnitrile, acmc-1cee4, benzonitrile,2,4,6-trimethyl
PubChem CID 137649
IUPAC Name 2,4,6-trimethylbenzonitrile
SMILES CC1=CC(=C(C(=C1)C)C#N)C

Specifications

Melting Point 50°C to 55°C
Boiling Point 79°C to 80°C (2 mmHg)
Quantity 25 g
UN Number UN3439
Beilstein 2044618
Solubility Information Slightly soluble in water.
Formula Weight 145.21
Percent Purity 98%
Chemical Name or Material 2,4,6-Trimethylbenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.