missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylbenzenesulfonyl hydrazide, 97%, Thermo Scientific™

Product Code. 10661642 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10661642 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10661642 Supplier Thermo Scientific Acros Supplier No. 185090250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 16182-15-3
MDL Number MFCD00007585
InChI Key JQUBKTQDNVZHIY-UHFFFAOYSA-N
Synonym 2,4,6-trimethylbenzenesulfonyl hydrazide, mesitylene-2-sulphonohydrazide, 2-mesitylenesulfonyl hydrazide, 2-mesitylenesulfonohydrazide, 2-mesitylenesulfono hydrazide, benzenesulfonic acid,2,4,6-trimethyl-, hydrazide, hydrazino 2,4,6-trimethylphenyl sulfone, mesitylenesulphonohydrazide, mesitylenesulfonylhydrazide, 2-mesitylehesulfonohydrazide
PubChem CID 85321
IUPAC Name 2,4,6-trimethylbenzenesulfonohydrazide
SMILES CC1=CC(=C(C(=C1)C)S(=O)(=O)NN)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 110.0°C to 114.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C9H14N2O2S
Linear Formula (CH3)3C6H2SO2NHNH2
Quantity 25 g
Molecular Weight (g/mol) 214.28
Formula Weight 214.28
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4, 6-Trimethylbenzenesulfonyl hydrazide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4,6-Trimethylbenzenesulfonyl hydrazide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H228-Flammable solid.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.