missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylaniline, 97%

Product Code. 10615212 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10615212 100 g Glass bottle 100g
10432845 500 g Glass bottle 500g
11955211 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10615212 Supplier Thermo Scientific Acros Supplier No. 157961000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 88-05-1 | C9H13N | 135.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-05-1
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00007740
InChI Key KWVPRPSXBZNOHS-UHFFFAOYSA-N
Synonym mesidine, aminomesitylene, mesitylamine, benzenamine, 2,4,6-trimethyl, mesidin, 2,4,6-trimethylbenzenamine, 2-aminomesitylene, 2-amino-1,3,5-trimethylbenzene, 2,4,6-trimethyl aniline, mesitylene, 2-amino
PubChem CID 6913
ChEBI CHEBI:82545
IUPAC Name 2,4,6-trimethylaniline
SMILES CC1=CC(=C(C(=C1)C)N)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -5.0°C
Density 0.9600g/mL
Boiling Point 233.0°C
Flash Point 96°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3C6H2NH2
Refractive Index 1.5505 to 1.5525
Quantity 100 g
Beilstein 12,116
Specific Gravity 0.96
Solubility Information Solubility in water: immiscible with water. Other solubilities: slightly soluble in carbon tetrachloride,soluble in most common solvents
Formula Weight 135.21
Percent Purity 97%
Chemical Name or Material 2, 4, 6-Trimethylaniline
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4,6-Trimethylaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.