missing translation for 'onlineSavingsMsg'
Learn More

2',4',6'-Trimethylacetophenone, 97+%

Product Code. 11434583
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434583 100 g 100g
11424583 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11434583 Supplier Thermo Scientific Alfa Aesar Supplier No. A11549.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1667-01-2 | C11H14O | 162.23 g/mol

2',4',6'-Trimethylacetophenone is used as laboratory chemicals, manufacture of substances and intermediated for chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′,4′,6′-Trimethylacetophenone is used as laboratory chemicals, manufacture of substances and intermediated for chemical synthesis.

Solubility
Difficult to mix.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1667-01-2
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.23
MDL Number MFCD00008735
InChI Key XWCIICLTKWRWCI-UHFFFAOYSA-N
Synonym 1-mesitylethanone, 2',4',6'-trimethylacetophenone, acetylmesitylene, acetomesitylene, mesityl methyl ketone, 1-2,4,6-trimethylphenyl ethanone, ethanone, 1-2,4,6-trimethylphenyl, 2-acetylmesitylene, 2,4,6-trimethylacetophenone, methyl 2,4,6-trimethylphenyl ketone
PubChem CID 15461
IUPAC Name 1-(2,4,6-trimethylphenyl)ethanone
SMILES CC(=O)C1=C(C)C=C(C)C=C1C

Specifications

Density 0.975
Boiling Point 235°C to 236°C
Flash Point >110°C (230°F)
Refractive Index 1.517
Quantity 100 g
Beilstein 2044595
Solubility Information Difficult to mix.
Formula Weight 162.23
Percent Purity 98%
Chemical Name or Material 2',4',6'-Trimethylacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.