missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethyl-N-[3-(trifluoromethyl)phenyl]benzenesulfonamide, Thermo Scientific™

Product Code. 15450487
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15450487 50 mg 50mg
15440487 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15450487 Supplier Thermo Scientific Alfa Aesar Supplier No. J65999.MF

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Activates phospholipase C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 200933-14-8
Molecular Formula C16H16F3NO2S
Molecular Weight (g/mol) 343.36
MDL Number MFCD00095824
InChI Key ZIIUUSVHCHPIQD-UHFFFAOYSA-N
Synonym m-3m3fbs, 2,4,6-trimethyl-n-3-trifluoromethyl phenyl benzenesulfonamide, m3m3fbs, 2,4,6-trimethyl-n-3-trifluoromethyl phenyl benzene-1-sulfonamide, 2,4,6-trimethyl-n-3-trifluoromethyl-phenyl-benzenesulfonamide, phospholipase c activator, m-3m3fbs, n-3-trifluoromethylphenyl-2,4,6-trimethylbenzenesulfonamide, 2,4,6-trimethyl-n-m-3-trifluoromethylphenyl benzenesulfonamide, tocris-1941
PubChem CID 761523
SMILES CC1=CC(C)=C(C(C)=C1)S(=O)(=O)NC1=CC=CC(=C1)C(F)(F)F

Specifications

Melting Point 114°C to 118°C
Color White
Quantity 50 mg
IUPAC Name 2,4,6-trimethyl-N-[3-(trifluoromethyl)phenyl]benzenesulfonamide
Formula Weight 343.36
Physical Form Powder
Chemical Name or Material 2,4,6-Trimethyl-N-[3-(trifluoromethyl)phenyl]benzenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.