missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number 10428530
Click to view available options
Quantity:
1 g
10 g
25 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
830-79-5
C10H12O4
196.2
MFCD00003313
CRBZVDLXAIFERF-UHFFFAOYSA-N
benzaldehyde, 2,4,6-trimethoxy, 2,4,6-trimethoxybenz aldehyde, pubchem8264, acmc-1cuhn, ksc490m2h, 2,4,6-trimethoxy-benzaldehyde, 2,4,6-trimethoxybenz-aldehyde, 2,4,6-trimethoxy benz aldehyde, crbzvdlxaiferf-uhfffaoysa, 1-formyl-2,4,6-trimethoxybenzene
70019
2,4,6-trimethoxybenzaldehyde
COC1=CC(=C(C(=C1)OC)C=O)OC
This item is not returnable. View return policy
830-79-5
C10H12O4
196.2
MFCD00003313
CRBZVDLXAIFERF-UHFFFAOYSA-N
benzaldehyde, 2,4,6-trimethoxy, 2,4,6-trimethoxybenz aldehyde, pubchem8264, acmc-1cuhn, ksc490m2h, 2,4,6-trimethoxy-benzaldehyde, 2,4,6-trimethoxybenz-aldehyde, 2,4,6-trimethoxy benz aldehyde, crbzvdlxaiferf-uhfffaoysa, 1-formyl-2,4,6-trimethoxybenzene
70019
2,4,6-trimethoxybenzaldehyde
COC1=CC(=C(C(=C1)OC)C=O)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 118.0°C to 120.0°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3O)3C6H2CHO
Quantity 10 g
Beilstein 08,39
Formula Weight 196.2
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4, 6-Trimethoxybenzaldehyde, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'