missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethoxybenzaldehyde, 98%

Product Code. 11453574
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453574 250 g 250g
11433574 10 g 10g
11443574 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11453574 Supplier Thermo Scientific Alfa Aesar Supplier No. A11238.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 830-79-5 | C10H12O4 | 196.202 g/mol

2,4,6-Trimethoxybenzaldehyde was used in the preparation and characterization of three RNA-specific fluorescent probes and their use in live cell imaging. It was used as starting reagent for the regioselective synthesis of new (±)-8-alkyl-5,7-dihydroxy-4-(4-hydroxyphenyl)-3,4-dihydrocoumarins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethoxybenzaldehyde was used in the preparation and characterization of three RNA-specific fluorescent probes and their use in live cell imaging. It was used as starting reagent for the regioselective synthesis of new (+/-)-8-alkyl-5,7-dihydroxy-4-(4-hydroxyphenyl)-3,4-dihydrocoumarins.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 830-79-5
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.202
MDL Number MFCD00003313
InChI Key CRBZVDLXAIFERF-UHFFFAOYSA-N
Synonym benzaldehyde, 2,4,6-trimethoxy, 2,4,6-trimethoxybenz aldehyde, pubchem8264, acmc-1cuhn, ksc490m2h, 2,4,6-trimethoxy-benzaldehyde, 2,4,6-trimethoxybenz-aldehyde, 2,4,6-trimethoxy benz aldehyde, crbzvdlxaiferf-uhfffaoysa, 1-formyl-2,4,6-trimethoxybenzene
PubChem CID 70019
IUPAC Name 2,4,6-trimethoxybenzaldehyde
SMILES COC1=CC(=C(C(=C1)OC)C=O)OC

Specifications

Melting Point 119°C to 123°C
Quantity 250 g
Beilstein 1956051
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 196.2
Percent Purity 98%
Chemical Name or Material 2,4,6-Trimethoxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.