missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethoxyaniline, 97%, Thermo Scientific™

Product Code. 10046132
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Glass/Safe-Cote
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 14227-17-9
Molecular Formula C9H13NO3
Molecular Weight (g/mol) 183.207
MDL Number MFCD00017165
InChI Key FNSAKXLEFPFZOM-UHFFFAOYSA-N
Synonym benzenamine, 2,4,6-trimethoxy, 2,4,6-tri-methoxyaniline, 2,4,6-trimethoxybenzenamine, 2,4,6-trimethoxy-phenylamine, 2,4,6-trimethoxy-phenyl-amine
PubChem CID 84271
IUPAC Name 2,4,6-trimethoxyaniline
SMILES COC1=CC(=C(C(=C1)OC)N)OC

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 183.21
Percent Purity 97%
Chemical Name or Material 2, 4, 6-Trimethoxyaniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'