missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triisopropylbenzoyl chloride, 98+%

Product Code. 11403644
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403644 5 g 5g
11413644 25 g 25g
11423644 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11403644 Supplier Thermo Scientific Alfa Aesar Supplier No. A11280.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 57199-00-5 | C16H23ClO | 266.809 g/mol

2,4,6-Triisopropylbenzoyl chloride is used as pharmaceuticcal intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Triisopropylbenzoyl chloride is used as pharmaceuticcal intermediates.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Store away from moisture and oxidizing agents. Incompatible with strong oxidizing agents, ammonia etc,.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57199-00-5
Molecular Formula C16H23ClO
Molecular Weight (g/mol) 266.809
MDL Number MFCD00015030
InChI Key OSKNTKJPGKHDHV-UHFFFAOYSA-N
Synonym 2,4,6-triisopropylbenzoyl chloride, benzoyl chloride, 2,4,6-tris 1-methylethyl, acmc-20aoev, 2,4,6-triiso-propylbenzoyl chloride, 2,4,6-tri propan-2-yl benzoyl chloride, benzoyl chloride,2,4,6-tris 1-methylethyl, 2,4,6-tris 1-methylethyl benzoic acid chloride
PubChem CID 92697
IUPAC Name 2,4,6-tri(propan-2-yl)benzoyl chloride
SMILES CC(C)C1=CC(=C(C(=C1)C(C)C)C(=O)Cl)C(C)C

Specifications

Melting Point 78°C to 82°C
Quantity 5 g
UN Number UN3261
Beilstein 2269534
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 266.81
Percent Purity ≥98%
Chemical Name or Material 2,4,6-Triisopropylbenzoyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.