missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triisopropylbenzoic acid, 97%

Product Code. 11452867
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11452867 50 g 50g
11442867 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11452867 Supplier Thermo Scientific Alfa Aesar Supplier No. A15489.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 49623-71-4 | C16H24O2 | 248.366 g/mol

2,4,6-Triisopropylbenzoic acid is a hindered acid intermediate compound in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Triisopropylbenzoic acid is a hindered acid intermediate compound in organic synthesis.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 49623-71-4
Molecular Formula C16H24O2
Molecular Weight (g/mol) 248.366
MDL Number MFCD00015031
InChI Key ULVHAZFBJJXIDO-UHFFFAOYSA-N
Synonym 2,4,6-triisopropylbenzoic acid, 2,4,6-triisopropyl benzoic acid, 2,4,6-tris 1-methylethyl benzoic acid, 2,4,6-tri propan-2-yl benzoic acid, benzoic acid,2,4,6-tris 1-methylethyl, benzoic acid, 2,4,6-tris 1-methylethyl, 2,4,6-tris methylethyl benzoic acid, acmc-20aoew, pubchem7877
PubChem CID 96214
IUPAC Name 2,4,6-tri(propan-2-yl)benzoic acid
SMILES CC(C)C1=CC(=C(C(=C1)C(C)C)C(=O)O)C(C)C

Specifications

Melting Point 184°C to 187°C
Quantity 50 g
Beilstein 2372331
Solubility Information Slightly soluble in water.
Formula Weight 248.37
Percent Purity 97%
Chemical Name or Material 2,4,6-Triisopropylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.