Learn More
2,4,6-Triisopropylbenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals
£33.20 - £501.00
Chemical Identifiers
CAS | 6553-96-4 |
---|---|
Molecular Formula | C15H23ClO2S |
Molecular Weight (g/mol) | 302.857 |
MDL Number | MFCD00007433 |
InChI Key | JAPYIBBSTJFDAK-UHFFFAOYSA-N |
Synonym | 2,4,6-triisopropylbenzenesulfonyl chloride, 2,4,6-triisopropylbenzene-1-sulfonyl chloride, trisyl chloride, 2,4,6-triisopropylbenzenesulphonyl chloride, benzenesulfonyl chloride, 2,4,6-tris 1-methylethyl, 2,4,6-triisopropylbenzenesulfonylchloride, 2,4,6-triisopropyl-benzenesulfonyl chloride, tpscl 2,4,6-triisopropyl benzenesulfonyl chloride, 2,4,6-triisopropylphenylsulfonyl chloride, 2,4,6-triisopropyl benzenesulfonyl chloride |
PubChem CID | 81042 |
IUPAC Name | 2,4,6-tri(propan-2-yl)benzenesulfonyl chloride |
SMILES | CC(C)C1=CC(=C(C(=C1)C(C)C)S(=O)(=O)Cl)C(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11450333
|
Thermo Scientific Alfa Aesar
A11458.14 |
25 g |
£33.20
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11460333
|
Thermo Scientific Alfa Aesar
A11458.22 |
100 g |
£115.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11470333
|
Thermo Scientific Alfa Aesar
A11458.36 |
500 g |
£501.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2,4,6-Triisopropylbenzenesulfonyl chloride is used for the synthesis of glycerophospholipids as a condensing agent and in the analysis of phosphonolipids in egg yolk by HPLC/MS technique. It acts as a coupling reagent for the synthesis of oligonucleotides and as sulfonates of pyrimidine nucleosides. It also serves as a condensing agent in the synthesis of hydrogen-phosphonate diesters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,4,6-Triisopropylbenzenesulfonyl chloride is used for the synthesis of glycerophospholipids as a condensing agent and in the analysis of phosphonolipids in egg yolk by HPLC/MS technique. It acts as a coupling reagent for the synthesis of oligonucleotides and as sulfonates of pyrimidine nucleosides. It also serves as a condensing agent in the synthesis of hydrogen-phosphonate diesters.
Solubility
Soluble in chloroform.
Notes
Incompatible with strong bases.
Chemical Identifiers
6553-96-4 | |
302.857 | |
JAPYIBBSTJFDAK-UHFFFAOYSA-N | |
81042 | |
CC(C)C1=CC(=C(C(=C1)C(C)C)S(=O)(=O)Cl)C(C)C |
C15H23ClO2S | |
MFCD00007433 | |
2,4,6-triisopropylbenzenesulfonyl chloride, 2,4,6-triisopropylbenzene-1-sulfonyl chloride, trisyl chloride, 2,4,6-triisopropylbenzenesulphonyl chloride, benzenesulfonyl chloride, 2,4,6-tris 1-methylethyl, 2,4,6-triisopropylbenzenesulfonylchloride, 2,4,6-triisopropyl-benzenesulfonyl chloride, tpscl 2,4,6-triisopropyl benzenesulfonyl chloride, 2,4,6-triisopropylphenylsulfonyl chloride, 2,4,6-triisopropyl benzenesulfonyl chloride | |
2,4,6-tri(propan-2-yl)benzenesulfonyl chloride |
Specifications
6553-96-4 | |
C15H23ClO2S | |
UN3261 | |
Moisture sensitive | |
Soluble in chloroform. | |
CC(C)C1=CC(=C(C(=C1)C(C)C)S(=O)(=O)Cl)C(C)C | |
302.857 | |
302.87 | |
2,4,6-Triisopropylbenzenesulfonyl chloride |
94°C to 97°C | |
MFCD00007433 | |
1218575 | |
2,4,6-triisopropylbenzenesulfonyl chloride, 2,4,6-triisopropylbenzene-1-sulfonyl chloride, trisyl chloride, 2,4,6-triisopropylbenzenesulphonyl chloride, benzenesulfonyl chloride, 2,4,6-tris 1-methylethyl, 2,4,6-triisopropylbenzenesulfonylchloride, 2,4,6-triisopropyl-benzenesulfonyl chloride, tpscl 2,4,6-triisopropyl benzenesulfonyl chloride, 2,4,6-triisopropylphenylsulfonyl chloride, 2,4,6-triisopropyl benzenesulfonyl chloride | |
JAPYIBBSTJFDAK-UHFFFAOYSA-N | |
2,4,6-tri(propan-2-yl)benzenesulfonyl chloride | |
81042 | |
98% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 229-479-6
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only