missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sorvall™ Legend™ T/RT Centrifuge Tube Adapters for Round Rotor Buckets

Product Code. 10202991 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Green
Light Gray
Olive Brown
Red
White
Yellow
Tube Size:
10/15 mL
15 mL
175/250 mL
50 mL
7 mL
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Tube Size unitSize
10202991 Red 15 mL Each
10290162 Yellow 7 mL Each
10782393 Light Gray 7 mL Each
10259301 White 10/15 mL Each
10384681 Olive Brown 15 mL Each
10395472 Green 50 mL Each
10181031 White 175/250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 10202991 Supplier Thermo Scientific™ Supplier No. 75008135
  • ONLINE EXCLUSIVE PRICE
    £153.00
    £143.37 / Pack of 1
    Save £9.63
    6% Off
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

For use with four-place rotors 75006445 and 75006434 when equipped with round bucket 75006441

Adapters sold individually; order correct quantity to equip your rotor.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) LH-4000W and LH-4000 rotor with round bucket
Color Red
Tube Size 15 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.