missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triisopropylbenzenesulfonyl azide, 98%, stab. with ca 10% water

Product Code. 11334316
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11334316

Brand: Thermo Scientific Alfa Aesar L19383.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 36982-84-0 | C15H23N3O2S | 309.428 g/mol

2,4,6-Triisopropylbenzenesulfonyl azide is used as a reagent for diazo transfer to ketones, e.g. under phase-transfer conditions, giving better yields of sterically-hindered ?-diazo ketones than other methods. It is also used for azide transfer to potassium enolates (KHMDS) to form the ?-azido derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Triisopropylbenzenesulfonyl azide is used as a reagent for diazo transfer to ketones, e.g. under phase-transfer conditions, giving better yields of sterically-hindered ɑ-diazo ketones than other methods. It is also used for azide transfer to potassium enolates (KHMDS) to form the ɑ-azido derivatives.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36982-84-0
Molecular Formula C15H23N3O2S
Molecular Weight (g/mol) 309.428
MDL Number MFCD00859286
InChI Key AEMWUHCKKDPRSK-UHFFFAOYSA-N
Synonym 2,4,6-triisopropylbenzenesulfonyl azide, 2,4,6-triisopropylbenzene-sulfonyl azide, trisyl azide, 2,4,6-triisopropylbenzene-sulfonyl azide,, benzenesulfonyl azide, 2,4,6-tris 1-methylethyl, 2,4,6-triisopropyl-benzenesulfonyl azide, n-diazo-2,4,6-triisopropyl-benzenesulfonamide, 2,4,6-tris 1-methylethyl-benzenesulfonyl azide, 2,4,6-triisopropylbenzenesulfonyl azide, stab. with ca water, trisyl azide solution
PubChem CID 371707
IUPAC Name N-diazo-2,4,6-tri(propan-2-yl)benzenesulfonamide
SMILES CC(C)C1=CC(=C(C(=C1)C(C)C)S(=O)(=O)N=[N+]=[N-])C(C)C

Specifications

Melting Point 40°C to 44°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 309.43
Percent Purity 98%
Chemical Name or Material 2,4,6-Triisopropylbenzenesulfonyl azide, stabilized with ≈10% water

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.