Learn More
2,4,6-Triisopropylbenzenesulfonyl azide, 98%, stab. with ca 10% water
Description
2,4,6-Triisopropylbenzenesulfonyl azide is used as a reagent for diazo transfer to ketones, e.g. under phase-transfer conditions, giving better yields of sterically-hindered ?-diazo ketones than other methods. It is also used for azide transfer to potassium enolates (KHMDS) to form the ?-azido derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,4,6-Triisopropylbenzenesulfonyl azide is used as a reagent for diazo transfer to ketones, e.g. under phase-transfer conditions, giving better yields of sterically-hindered ɑ-diazo ketones than other methods. It is also used for azide transfer to potassium enolates (KHMDS) to form the ɑ-azido derivatives.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C.
Specifications
Specifications
| Melting Point | 40°C to 44°C |
| Quantity | 1 g |
| Solubility Information | Insoluble in water. |
| Formula Weight | 309.43 |
| Percent Purity | 98% |
| Chemical Name or Material | 2,4,6-Triisopropylbenzenesulfonyl azide, stabilized with ≈10% water |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.