missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triisopropylbenzeneboronic acid, 98%

Product Code. 11409947
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409947 1 g 1g
11419947 5 g 5g
11429947 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11409947 Supplier Thermo Scientific Alfa Aesar Supplier No. B22891.03
 Restricted Item
Estimated Shipment: 16-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 154549-38-9 | C15H25BO2 | 248.173 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154549-38-9
Molecular Formula C15H25BO2
Molecular Weight (g/mol) 248.173
MDL Number MFCD02683099
InChI Key FGFYEKLWZBTLEW-UHFFFAOYSA-N
Synonym 2,4,6-triisopropylphenyl boronic acid, 2,4,6-triisopropylphenylboronic acid, 2,4,6-triisopropylbenzeneboronic acid, 2,4,6-tri propan-2-yl phenyl boronic acid, 2,4,6-tris propan-2-yl phenyl boronic acid, acmc-209dav, 2,4,6-triisopropylphenyiboronic acid, 2,4,6-triisopropylphenyl boronicacid
PubChem CID 15153544
IUPAC Name [2,4,6-tri(propan-2-yl)phenyl]boronic acid
SMILES B(C1=C(C=C(C=C1C(C)C)C(C)C)C(C)C)(O)O

Specifications

Melting Point ∼165°C (decomposition)
Quantity 1 g
Formula Weight 248.16
Percent Purity 98%
Chemical Name or Material 2,4,6-Triisopropylbenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.