missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trifluorobenzoyl chloride, 97%

Product Code. 11449846
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11449846

Brand: Thermo Scientific Alfa Aesar B20323.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 79538-29-7 | C7H2ClF3O | 194.537 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79538-29-7
Molecular Formula C7H2ClF3O
Molecular Weight (g/mol) 194.537
MDL Number MFCD00070810
InChI Key SIFIJQFBERMWMU-UHFFFAOYSA-N
Synonym 2,4,6-trifluorobenzoylchloride, 2,4,6-trifluoro-benzoyl chloride, acmc-20aof3, timtec-bb sbb005827, 2,4,6-tri-fluorobenzoyl chloride, 2,4,6-trifluorobenzoyl chloride, benzoyl chloride,2,4,6-trifluoro, benzoyl chloride, 2,4,6-trifluoro, 2,4,6-trifluorobenzoyl chloride 1g, benzoyl chloride, 2,4,6-trifluoro-9ci
PubChem CID 144895
IUPAC Name 2,4,6-trifluorobenzoyl chloride
SMILES C1=C(C=C(C(=C1F)C(=O)Cl)F)F

Specifications

Density 1.499
Boiling Point 167°C to 168°C
Refractive Index 1.484
Quantity 1 g
UN Number UN1760
Beilstein 1958563
Sensitivity Moisture sensitive
Formula Weight 194.54
Percent Purity 97%
Chemical Name or Material 2,4,6-Trifluorobenzoyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.