missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trifluorobenzotrifluoride, 96%

Product Code. 11394925
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394925 1 g 1g
11314935 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394925 Supplier Thermo Scientific Alfa Aesar Supplier No. L18167.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 122030-04-0 | C7H2F6 | 200.083 g/mol

2,4,6-Trifluorobenzotrifluoride finds its application as an intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trifluorobenzotrifluoride finds its application as an intermediate in organic syntheses.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away. Protect from electrostatic charges.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122030-04-0
Molecular Formula C7H2F6
Molecular Weight (g/mol) 200.083
MDL Number MFCD00190109
InChI Key SGPXFVDJRSZIPK-UHFFFAOYSA-N
Synonym 2,4,6-trifluorobenzotrifluoride, 1,3,5-trifluoro-2-trifluoromethyl benzene, pubchem4877, 2,4,6-trifluorobenzotrifluorode, 2,4,6-tri-fluorobenzotrifluoride, benzene, 1,3,5-trifluoro-2-trifluoromethyl, alpha,alpha,alpha,2,4,6-hexafluorotoluene, alpha,alpha,alpha,2,4,6-hexafluorotoluene;1,3,5-trifluoro-2-trifluoromethyl benzene
PubChem CID 2734030
IUPAC Name 1,3,5-trifluoro-2-(trifluoromethyl)benzene
SMILES C1=C(C=C(C(=C1F)C(F)(F)F)F)F

Specifications

Density 1.475
Boiling Point 105°C to 107°C
Flash Point 22°C (71°F)
Refractive Index 1.385
Quantity 1 g
UN Number UN1993
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 200.08
Percent Purity 99%
Chemical Name or Material 2,4,6-Trifluorobenzotrifluoride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.