missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trifluorobenzoic acid, 98%

Product Code. 11448637
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11448637

Brand: Thermo Scientific Alfa Aesar A19758.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 28314-80-9 | C7H3F3O2 | 176.094 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28314-80-9
Molecular Formula C7H3F3O2
Molecular Weight (g/mol) 176.094
MDL Number MFCD00042398
InChI Key SJZATRRXUILGHH-UHFFFAOYSA-N
Synonym benzoic acid, 2,4,6-trifluoro, 2,4,6-trifluorobenzoicacid, the derivatives, 2,4,6-trifluorobenzioc acid, pubchem1347, acmc-209h1r, ksc204g9t, 2,4,6-trifluoro-benzoic acid, rarechem al bo 0448, benzoicacid, 2,4,6-trifluoro
PubChem CID 520373
IUPAC Name 2,4,6-trifluorobenzoic acid
SMILES C1=C(C=C(C(=C1F)C(=O)O)F)F

Specifications

Melting Point 140°C to 142°C
Quantity 1 g
Beilstein 1958300
Formula Weight 176.1
Percent Purity 98%
Chemical Name or Material 2,4,6-Trifluorobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.