missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trichloropyridine, 98%

Product Code. 10254634 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10254634 5 g Glass bottle 5g
10336884 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10254634 Supplier Thermo Scientific Acros Supplier No. 399590050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16063-69-7 | C5H2Cl3N | 182.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16063-69-7
Molecular Formula C5H2Cl3N
Molecular Weight (g/mol) 182.44
MDL Number MFCD00971928
InChI Key FJNNGKMAGDPVIU-UHFFFAOYSA-N
Synonym pyridine, 2,4,6-trichloro, 2,4,6-trichloro-pyridine, pubchem11020, acmc-1bu24, ksc497g8t, 2,4,6-tris chloranyl pyridine, 2,4,6-trichloropyridine
PubChem CID 27665
IUPAC Name 2,4,6-trichloropyridine
SMILES C1=C(C=C(N=C1Cl)Cl)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 33°C to 37°C
Color Brown-Yellow to Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 182.44
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4, 6-Trichloropyridine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4,6-Trichloropyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.