missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trichloropyridine, 97%

Product Code. p-7037937
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15443328

Brand: Thermo Scientific Alfa Aesar H56787.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16063-69-7 | C5H2Cl3N | 182.428 g/mol

It is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediates.

Solubility
Sparingly soluble in water. (0.113 mm Hg at 25°C)

Notes
Store at 2-8°C. Avoid using oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16063-69-7
Molecular Formula C5H2Cl3N
Molecular Weight (g/mol) 182.428
MDL Number MFCD00971928
InChI Key FJNNGKMAGDPVIU-UHFFFAOYSA-N
Synonym pyridine, 2,4,6-trichloro, 2,4,6-trichloro-pyridine, pubchem11020, acmc-1bu24, ksc497g8t, 2,4,6-tris chloranyl pyridine, 2,4,6-trichloropyridine
PubChem CID 27665
IUPAC Name 2,4,6-trichloropyridine
SMILES C1=C(C=C(N=C1Cl)Cl)Cl

Specifications

Melting Point 33°C to 37°C
Flash Point >110°C (230°F)
Quantity 5 g
Solubility Information Sparingly soluble in water. (0.113 mm Hg at 25°C)
Formula Weight 182.44
Percent Purity 97%
Chemical Name or Material 2,4,6-Trichloropyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.