missing translation for 'onlineSavingsMsg'
Learn More

(2,4,6-Trichlorophenoxy)acetic Acid, TRC

Product Code. 30452844
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30452844 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30452844 Supplier Toronto Research Chemicals Supplier No. TRCT774580500MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 575-89-3
Molecular Formula C8 H5 Cl3 O3
Quantity 500 mg
Synonym 2,4,6, Trichlorophenoxyacetic acid (BTS 9608), Acetic acid, 2-(2,4,6-trichlorophenoxy)- (ACI), 2-(2,4,6-Trichlorophenoxy)acetic acid (ACI), Acetic acid, (2,4,6-trichlorophenoxy)- (8CI, 9CI), (2,4,6-Trichlorophenoxy)acetic acid, 2,4,6-T, NSC 61993
InChI Formula InChI=1S/C8H5Cl3O3/c9-4-1-5(10)8(6(11)2-4)14-3-7(12)13/h1-2H,3H2,(H,12,13)
SMILES OC(=O)COc1c(Cl)cc(Cl)cc1Cl
IUPAC Name 2-(2,4,6-trichlorophenoxy)acetic acid
Molecular Weight (g/mol) 255.48
Formula Weight 253.9304
Chemical Name or Material 2,4,6-Trichlorophenoxyacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.