missing translation for 'onlineSavingsMsg'
Learn More

(2,4,6-Trichlorophenoxy)acetic Acid-13C6, TRC

Product Code. 30449263
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30449263 1 mg 1mg
30452440 10 mg 10mg
30449681 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30449263 Supplier Toronto Research Chemicals Supplier No. TRCT7745821MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2,4,6-Trichlorophenoxy)acetic Acid-13C6
Molecular Formula C213C6H5Cl3O3
Quantity 1 mg
Synonym 2-(2,4,6-Trichlorophenoxy)acetic Acid-13C6, 2,4,6-T-13C6, NSC 61993-13C6
InChI Formula InChI=1S/C8H5Cl3O3/c9-4-1-5(10)8(6(11)2-4)14-3-7(12)13/h1-2H,3H2,(H,12,13)/i1+1,2+1,4+1,5+1,6+1,8+1
SMILES O=C(O)CO[13C]1=[13C](Cl)[13CH]=[13C](Cl)[13CH]=[13C]1Cl
Molecular Weight (g/mol) 261.44
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.