missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trichlorobenzoyl chloride, 98%

Product Code. 11924881 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11924881 25 g Glass bottle 25g
10645541 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11924881 Supplier Thermo Scientific Acros Supplier No. 335180250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4136-95-2 | C7H2Cl4O | 243.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4136-95-2
Molecular Formula C7H2Cl4O
Molecular Weight (g/mol) 243.89
MDL Number MFCD00075323
InChI Key OZGSEIVTQLXWRO-UHFFFAOYSA-N
Synonym 2,4,6-trichlorobenzoylchloride, benzoyl chloride, 2,4,6-trichloro, mi, acmc-209jk3, ksc494s2p, 2,4,6-cl3c6h2cocl, 2,4,6-ttrichlorobenzoyl chloride, 2,4,6-trichloro benzoyl chloride, 2,4,6-trichlorobenzoic acid chloride, 2,4,6-tris chloranyl benzoyl chloride
PubChem CID 2733703
IUPAC Name 2,4,6-trichlorobenzoyl chloride
SMILES ClC(=O)C1=C(Cl)C=C(Cl)C=C1Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.5600g/mL
Boiling Point 107.0°C to 108.0°C (6.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5750 to 1.5770
Quantity 25 g
Merck Index 15,9798
Specific Gravity 1.56
Formula Weight 243.9
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 4, 6-Trichlorobenzoyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 2,4,6-Trichlorobenzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.