missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Trichlorobenzoyl chloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
2,4,6-Trichlorobenzoyl chloride is used in the preparation of γ-lactone and δ-lactone, aliphatic aromatic anhydrides, required for the synthesis of amphiphilic hyaluronan, mixed anhydride, required for the synthesis of angelate esters, synthesis of both spongistatin 1 and spongistatin 2 and large-ring lactones in high yields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,4,6-Trichlorobenzoyl chloride is used in the preparation of γ-lactone and δ-lactone, aliphatic aromatic anhydrides, required for the synthesis of amphiphilic hyaluronan, mixed anhydride, required for the synthesis of angelate esters, synthesis of both spongistatin 1 and spongistatin 2 and large-ring lactones in high yields.
Solubility
Reacts with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive to moisture. Incompatible with strong bases and strong oxidizing agents.
2,4,6-Trichlorobenzoyl chloride is used in the preparation of γ-lactone and δ-lactone, aliphatic aromatic anhydrides, required for the synthesis of amphiphilic hyaluronan, mixed anhydride, required for the synthesis of angelate esters, synthesis of both spongistatin 1 and spongistatin 2 and large-ring lactones in high yields.
Solubility
Reacts with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive to moisture. Incompatible with strong bases and strong oxidizing agents.
Specifications
Density | 1.561 |
Boiling Point | 107°C to 108°C (6 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.575 |
Quantity | 5 g |
UN Number | UN3265 |
Beilstein | 2050280 |
Sensitivity | Moisture sensitive |
Solubility Information | Reacts with water. |
Formula Weight | 243.91 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'