missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tribromotoluene, 98+%

Product Code. 11314484
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11314484 10 g 10g
11324484 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11314484 Supplier Thermo Scientific Alfa Aesar Supplier No. L01149.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6320-40-7 | C7H5Br3 | 328.829 g/mol

2,4,6-Tribromotoluene, is used as a chemical and organic intermediate. It can be used to produce 2,4,6-tribromo-benzyl bromide at temperature of 200 - 220°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Tribromotoluene, is used as a chemical and organic intermediate. It can be used to produce 2,4,6-tribromo-benzyl bromide at temperature of 200 - 220°C.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6320-40-7
Molecular Formula C7H5Br3
Molecular Weight (g/mol) 328.829
MDL Number MFCD00013527
InChI Key BFRIZWKDNUHPHL-UHFFFAOYSA-N
Synonym 2,4,6-tribromotoluene, benzene, 1,3,5-tribromo-2-methyl, acmc-20aof7, 2,4,6,tribromotoluene, 2,4,6-tribrom-toluol, 2-methyl-1,3,5-tribromobenzene, benzene,1,3,5-tribromo-2-methyl, 2,4,6-tribromotoluene 10g, 1,3,5-tribromo-2-methyl-benzene
PubChem CID 33916
IUPAC Name 1,3,5-tribromo-2-methylbenzene
SMILES CC1=C(C=C(C=C1Br)Br)Br

Specifications

Melting Point 67°C to 70°C
Density 2.479
Boiling Point 290°C
Quantity 10 g
Beilstein 2359313
Solubility Information Insoluble in water.
Formula Weight 328.83
Percent Purity ≥98%
Chemical Name or Material 2,4,6-Tribromotoluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.