missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tribromophenyl Beta-D-Glucopyranoside, TRC

Product Code. 30455897
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455897 25 mg 25mg
30456969 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30455897 Supplier Toronto Research Chemicals Supplier No. TRCT77267025MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 502495-87-6
Molecular Formula C12H13Br3O6
Molecular Weight (g/mol) 492.94
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(2,4,6-tribromophenoxy)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@H](Oc2c(Br)cc(Br)cc2Br)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C12H13Br3O6/c13-4-1-5(14)11(6(15)2-4)21-12-10(19)9(18)8(17)7(3-16)20-12/h1-2,7-10,12,16-19H,3H2/t7-,8-,9+,10-,12+/m1/s1
Formula Weight 489.8262
Chemical Name or Material 2,4,6-Tribromophenyl beta-D-Glucopyranoside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.