missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tribromophenol, Mikromol™

Codice prodotto. 30460087
Cambia vista
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Dimensione della confezione:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Dimensione della confezione
30460087 4 x 25 mg -
17951006 25 mg 25mg
17914066 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. View return policy
Product Code. 30460087 Supplier Mikromol Supplier No. MM1200.0500254AMP
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2,4,6-Tribromophenol, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4,6-Tribromophenol
CAS 118-79-6
Analyte or Component Names 2,4,6-Tribromophenol
Shipping Condition Room Temperature
Molecular Formula C6 H3 Br3 O
Certifications/Compliance ISO 17025
Synonym 2,4,6-Tribromophenol, Dembrexine Hydrochloride Monohydrate Imp. E (EP), Dembrexine Imp. E (EP), Dembrexine Hydrochloride Monohydrate Imp. E (EP), Phenol, 2,4,6-tribromo-, 1,3,5-Tribromo-2-hydroxybenzene, Bromkal Pur 3, Bromol, Flammex 3BP, NSC 2136, PH 73, Dembrexine Hydrochloride Monohydrate Impurity E, Dembrexine Impurity E
InChI Formula InChI=1S/C6H3Br3O/c7-3-1-4(8)6(10)5(9)2-3/h1-2,10H
Solution Type Neat
SMILES Oc1c(Br)cc(Br)cc1Br
IUPAC Name 2,4,6-tribromophenol
Molecular Weight (g/mol) 330.8
Quantity 4 x 25 mg
Formula Weight 327.7734 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.