missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tribromophenol, Mikromol™

Product Code. 17951006
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 17951006

Brand: Mikromol MM1200.050025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2,4,6-Tribromophenol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4,6-Tribromophenol
CAS 118-79-6
Analyte or Component Names 2,4,6-Tribromophenol
Shipping Condition Room Temperature
Molecular Formula C6 H3 Br3 O
Certifications/Compliance ISO 17025
Synonym 2,4,6-Tribromophenol, Dembrexine Hydrochloride Monohydrate Imp. E (EP), Dembrexine Imp. E (EP), Dembrexine Hydrochloride Monohydrate Imp. E (EP), Phenol, 2,4,6-tribromo-, 1,3,5-Tribromo-2-hydroxybenzene, Bromkal Pur 3, Bromol, Flammex 3BP, NSC 2136, PH 73, Dembrexine Hydrochloride Monohydrate Impurity E, Dembrexine Impurity E
InChI Formula InChI=1S/C6H3Br3O/c7-3-1-4(8)6(10)5(9)2-3/h1-2,10H
SMILES Oc1c(Br)cc(Br)cc1Br
IUPAC Name 2,4,6-tribromophenol
Molecular Weight (g/mol) 330.8
Quantity 25 mg
Formula Weight 327.7734 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.