missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tribromophenol, 98%

Product Code. 11429623
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429623 1000 g 1000g
11439623 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429623 Supplier Thermo Scientific Alfa Aesar Supplier No. A12424.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 118-79-6 | C6H3Br3O | 330.80 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-79-6
Molecular Formula C6H3Br3O
Molecular Weight (g/mol) 330.80
MDL Number MFCD00002150
InChI Key BSWWXRFVMJHFBN-UHFFFAOYSA-N
Synonym tribromophenol, bromol, bromkal pur 3, xeroform, phenol, 2,4,6-tribromo, flammex 3bp, unii-ys6k3eu393, ccris 1658, great lakes ph-73, 5175-83-7 bismuth 3+ salt
PubChem CID 1483
ChEBI CHEBI:47696
IUPAC Name 2,4,6-tribromophenol
SMILES OC1=C(Br)C=C(Br)C=C1Br

Specifications

Melting Point 86°C to 92°C
Density 2.55
Boiling Point 282°C to 290°C (sublimation)
Quantity 1000 g
UN Number UN3077
Beilstein 776920
Merck Index 14,9613
Formula Weight 330.8
Percent Purity 98%
Chemical Name or Material 2,4,6-Tribromophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.